missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Mycophenolate mofetil
GREENER_CHOICE

Product Code. 18073922
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18073922 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18073922 Supplier Tocris Bioscience™ Supplier No. 4102/50
  • ONLINE EXCLUSIVE PRICE
    123.00 €
    115.50 € / 50mg
    Save 7.50 €
    6% Off
 Restricted Item
Estimated Shipment: 28-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Inosine monophosphate dehydrogenase inhibitor

Immunosuppressant prodrug of mycophenolic acid (MPA) (Cat. No. 1505). Hydrolyzed by esterases to release MPA (a reversible inhibitor of inosine monophosphate dehydrogenase). Used to prevent organ rejection after transplantation. Displays inhibitory activity of lymphocyte functions. Inhibits type 1 collagen expression, enhances MMP-1 expression, and reduces α-smooth muscle actin gene expression.
TRUSTED_SUSTAINABILITY

Specifications

CAS 128794-94-5
Quantity 50 mg
Molecular Formula C23H31NO7
Formula Weight 433.49
Purity >99%
Molecular Weight (g/mol) 433.501
Chemical Name or Material Mycophenolate mofetil
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.