missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mycophenolic acid

Product Code. 18712604
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18712604 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18712604 Supplier R&D Systems™ Supplier No. 1505/100
  • ONLINE EXCLUSIVE PRICE
    154.49 €
    139.65 € / 100mg
    Save 14.84 €
    9% Off
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Inosine monophosphatase dehydrogenase inhibitor

Immunosuppressive agent with antiviral and antitumor effects in vitro and in vivo. Potently inhibits inosine monophosphate dehydrogenase, thus inhibiting de novo GTP synthesis leading to decreased RNA and DNA synthesis. Reversibly inhibits proliferation of T and B lymphocytes and antibody formation.
TRUSTED_SUSTAINABILITY

Specifications

CAS 24280-93-1
Quantity 100 mg
Product Line Tocris
Molecular Formula C17H20O6
Formula Weight 320.34
Purity >98%
Molecular Weight (g/mol) 320.341
Chemical Name or Material Mycophenolic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.