missing translation for 'onlineSavingsMsg'
Learn More

Mycophenolic acid, 98%

Product Code. p-7042263
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
This item is not returnable. View return policy

Product Code. 15414399

Brand: Thermo Scientific Alfa Aesar J61905.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 24280-93-1 | C17H20O6 | 320.341 g/mol

It is used as a small molecule IMPDH inhibitor. mycophenolic Acid is used as an immunosuppressive agent with antiviral and antitumor effects. It potently inhibits inosine monophosphate dehydrogenase, thus inhibiting de novo GTP synthesis leading to decreased RNA and DNA synthesis. It also reversibly inhibits proliferation of T and B lymphocytes and antibody formation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a small molecule IMPDH inhibitor. mycophenolic Acid is used as an immunosuppressive agent with antiviral and antitumor effects. It potently inhibits inosine monophosphate dehydrogenase, thus inhibiting de novo GTP synthesis leading to decreased RNA and DNA synthesis. It also reversibly inhibits proliferation of T and B lymphocytes and antibody formation.

Solubility
Soluble in methanol at 50mg/ml

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: 2 - 8°C. Keep away from strong oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24280-93-1
Molecular Formula C17H20O6
Molecular Weight (g/mol) 320.341
MDL Number MFCD00036814
InChI Key HPNSFSBZBAHARI-RUDMXATFSA-N
Synonym mycophenolic acid, mycophenolate, melbex, myfortic, mycophenolsaeure, acido micofenolico, micofenolico acido, acide mycophenolique, acidum mycophenolicum, 6-4-hydroxy-6-methoxy-7-methyl-3-oxo-5-phthalanyl-4-methyl-4-hexenoic acid
PubChem CID 446541
ChEBI CHEBI:168396
IUPAC Name (E)-6-(4-hydroxy-6-methoxy-7-methyl-3-oxo-1H-2-benzofuran-5-yl)-4-methylhex-4-enoic acid
SMILES CC1=C(C(=C(C2=C1COC2=O)O)CC=C(C)CCC(=O)O)OC

Specifications

Melting Point 139°C to 141°C
Color White
Quantity 1 g
Beilstein 1295848
Merck Index 14,6327
Solubility Information Soluble in methanol at 50mg/ml
Formula Weight 320.34
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Mycophenolic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.