missing translation for 'onlineSavingsMsg'
Learn More

Mycophenolic Acid Beta-D-Glucuronide, TRC

Product Code. 30011090
Change view
Click to view available options
Quantity:
1 mg
2.5 mg
10 mg
Unit Size:
10mg
1mg
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30011090 10 mg 10mg
30006834 1 mg 1mg
30007823 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30011090 Supplier Toronto Research Chemicals Supplier No. TRCM83152010MG
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 31528-44-6
Molecular Formula C23 H28 O12
Molecular Weight (g/mol) 496.46
Synonym Mycophenolic Acid β-D-Glucuronide
IUPAC Name (2S,3S,4S,5R,6S)-6-[[5-[(E)-5-carboxy-3-methylpent-2-enyl]-6-methoxy-7-methyl-3-oxo-1H-2-benzofuran-4-yl]oxy]-3,4,5-trihydroxyoxane-2-carboxylic acid
SMILES COc1c(C)c2COC(=O)c2c(O[C@@H]3O[C@@H]([C@@H](O)[C@H](O)[C@H]3O)C(=O)O)c1C\C=C(/C)\CCC(=O)O

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C23H28O12/c1-9(5-7-13(24)25)4-6-11-18(32-3)10(2)12-8-33-22(31)14(12)19(11)34-23-17(28)15(26)16(27)20(35-23)21(29)30/h4,15-17,20,23,26-28H,5-8H2,1-3H3,(H,24,25)(H,29,30)/b9-4+/t15-,16-,17+,20-,23+/m0/s1
Formula Weight 496.1581
Percent Purity >95%
Chemical Name or Material Mycophenolic acid-beta-D-glucuronide
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.