missing translation for 'onlineSavingsMsg'
Learn More

TOKU-E™ Mycophenolic Acid ReadyMade™ Solution

Product Code. 30044035
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 1 mL
Unit Size:
10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30044035 10 x 1 mL 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30044035 Supplier TOKU-E™ Supplier No. M12110x1ML
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Mycophenolic Acid ReadyMade™ Solution is provided as a sterile-filtered solution in a solution of β-cyclodextrin.

  • CAS Number: 24280-93-1
  • Molecular Formula: C17H20O6
  • Molecular Weight: 320.34 g/mol
  • Storage Conditions: -20°C
  • Form: Liquid
  • Source: Penicillium stoloniferum
  • Spectrum: Anisomycin is active against Gram-negative bacteria and protozoa. It has in vitro activity against dengue and Zika flaviviruses in cell culture
  • Mechanism of Action: Mycophenolic Acid blocks inosine monophosphate dehydrogenase in the guanosine monophosphate pathway and suppresses cytokine-induced nitric oxide production

Specifications

Chemical Name or Material Mycophenolic Acid
CAS 24280-93-1
Quantity 10 x 1 mL
Shipping Condition Ice Pack
Product Line ReadyMade™ Solutions
Target Iosine monophosphate dehydrogenase
Molecular Formula C17H20O6
Formula Weight 320.34 amu
Physical Form Liquid
Color Colorless
Product Type Antibiotic
Purity >98%
Molecular Weight (g/mol) 320.34 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.