missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Mycoplasma Hoechst Stain Kit

Product Code. 11495065 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 Tests
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11495065 100 Tests Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11495065 Supplier MP Biomedicals™ Supplier No. 093030000

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The Mycoplasma Hoechst Stain Kit is designed for rapid in situ detection of mycoplasma, and other prokaryotic organisms, in cell cultures in less than two hours.

  • Uses DNA fluorochrome staining technology with Hoechst Stain 33258
  • For the in situ detection of mycoplasma, bacteria, yeast, and fungi in cell cultures
  • Testing is rapid (less than 2 hours)and economical (100 tests per kit)
  • Non-nuclear staining is readily apparent, with contaminants standing out boldly against a black background

Specifications

Content And Storage 15° to 30°C
Product Type Mycoplasma Detection
Quantity 100 Tests

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.