missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ MycoSEQ™ Plus Mycoplasma Detection Kits

Product Code. 18755455 Shop All Applied Biosystems Products
Change view
Click to view available options
Includes:
Mycoplasma Detection Kit Only
PrepSEQ™ 1-2-3 Nucleic Acid Extraction Kit
PrepSEQ™ Express Nucleic Acid Extraction Kit
Unit Size:
100 reactions
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Includes Unit Size
18755455 PrepSEQ™ Express Nucleic Acid Extraction Kit 100 reactions
18765485 Mycoplasma Detection Kit Only Each
18872368 Mycoplasma Detection Kit Only Each
18775505 PrepSEQ™ 1-2-3 Nucleic Acid Extraction Kit Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 18755455 Supplier Applied Biosystems™ Supplier No. A57926
  • ONLINE EXCLUSIVE PRICE
    5630.00 €
    4870.00 € / 100 reactions
    Save 760.00 €
    13% Off
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The MycoSEQ™ Plus Mycoplasma Detection Kit is a TaqMan™-based quantitative PCR (qPCR) kit for the detection of mycoplasma in complex bioproduction samples.

The MycoSEQ™ Plus Mycoplasma Detection Kit is a TaqMan™-based quantitative PCR (qPCR) kit for the detection of mycoplasma in complex bioproduction samples. Designed and tested using criteria often applied toward rapid mycoplasma detection in biotherapeutic manufacturing cell culture lot release, the MycoSEQ Plus kit enables results that meet or exceed guidance on sensitivity and specificity expectations as described in European Pharmacopoeia (E.P. 2.6.7, 2007), US Pharmacopoeia (US63) and Japanese Pharmacopoeia. When used with a suitable sample preparation method, the MycoSEQ Plus kit can detect less than 10 CFU/mL or the genomic equivalent of 10 GC/mL.

This multiplexed assay enables predicted simultaneous screening of over 200 species of Mycoplasma target, discriminatory detection of an assay specific control template, and internal PCR control with no cross-reactivity to closely-related bacterial species.

Features of the MycoSEQ Plus Mycoplasma Detection Kit

  • Provides results in less than 1 day
  • Includes a unique discriminatory positive control to differentiate a true positive result from a potential cross-contamination positive call
  • Includes protocols for various methods
  • Live mycoplasma is not needed: MycoSEQ Mycoplasma Detection Kit does not contain any live mycoplasma and does not require use of live mycoplasma for validation
  • Can be used with gDNA, inactivated mycoplasma or live mycoplasma
  • May be used as an alternative to the standard 28-day culture test
  • May be used in conjunction with culture-based methods by providing preliminary results while waiting for results from the 28-day test

PrepSEQ nucleic acid extraction kits offer a robust and easy workflow to overcome complex matrix challenges to ensure the required sensitivity. Use MycoSEQ Plus Mycoplasma Detection Kit with PrepSEQ 1-2-3 Nucleic Acid Extraction Kit (Cat. No. A57925) for manual extractions, or MycoSEQ Plus Mycoplasma Detection Kit with PrepSEQ Express Nucleic Acid Extraction Kit (Cat. No. A57926) for automated extraction using the Automate Express Nucleic Acid Extraction System.

Integrated workflow solution

The MycoSEQ Plus Mycoplasma Detection Kit is part of an integrated workflow for adventitious agent, impurity and contaminant testing during biopharmaceutical manufacturing. The entire workflow including PrepSEQ Sample Preparation Kits, which offer manual or automated sample preparation approaches, together with qPCR analysis can provide results in under 5 hours. (Please note complex matrices may require additional upfront processing steps, as described in various available protocols.)

MycoSEQ Plus kit performance has been tested and confirmed on the Applied Biosystems 7500 Fast Real-Time PCR System and the Applied Biosystems QuantStudio 5 Real-Time PCR System. Data analysis using AccuSEQ Real-Time PCR Detection Software provides accurate quantitation and security, audit, and e-signature capabilities to help enable 21 CFR Pt 11 compliance.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage MycoSEQ Mycoplasma Detection Kit
• 2X qPCR Master Mix Plus, -15 to -25°C
• MycoSEQ Plus 10X Assay Mix, -15 to -25°C
• Negative Control, -15 to -25°C until first thawed, then store at 2°C to 8°C
• DNA Dilution Buffer, -15 to -25°C until first thawed, then store at 2°C to 8°C
• MycoSEQ Plus Discriminatory Positive Control, -15 to -25°C

PrepSEQ Express Nucleic Acid Sample Extraction Kit
• PrepSEQ Express Cartidges, store at 18-25°C
• Lysis Tubes, store at 18-25°C
• Sample Tubes, store at 18-25°C
• Elution Buffer, store at 18-25°C
• Elution Tubes, store at 18-25°C
• AutoMate Exptress Tips and Tip Holders, store at 18-25°C
Description MycoSEQ™ Plus Mycoplasma Detection Kit with PrepSEQ™ Express Nucleic Acid Extraction Kit
Detection Method Primer-probe
Form Liquid
GC-Rich PCR Performance Medium
PCR Method Multiplex qPCR
Reaction Speed Standard
For Use With (Equipment) QS5 0.1 mL Block or 7500 FAST
Label or Dye FAM
Product Line MycoSEQ
Product Type Mycoplasma Detection
Quantity 100 Reactions
Shipping Condition Dry Ice
Target Organism Class Mollicutes, mycoplasma
For Use With (Application) Microbial Detection
No. of Reactions 100 Reactions
Sample Type Biopharmaceutical Purification Sample, Bulk Drug Substance Sample, Cell Cultures
Includes PrepSEQ™ Express Nucleic Acid Extraction Kit
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.