Learn More
Lonza™ MycoZap™ Mycoplasma Elimination Reagent
Product Code. 11610281
Change view
Click to view available options
Quantity:
1 x 0.5 mL MycoZap™ Reagent 1, 1 x 1.5 mL MycoZap™ Reagent 2
1 x 2.5 mL MycoZap™ Reagent 1, 1 x 7.5 mL MycoZap™ Reagent 2
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11610281 1 x 0.5 mL MycoZap™ Reagent 1, 1 x 1.5 mL MycoZap™ Reagent 2 Each
11620281 1 x 2.5 mL MycoZap™ Reagent 1, 1 x 7.5 mL MycoZap™ Reagent 2 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11610281 Supplier Lonza™ Supplier No. LT07818
 Restricted Item
Estimated Shipment: 23-07-2026
Add to basket
Add to basket
Antibiotic formulation to remove Mycoplasma from contaminated cell cultures
  • Efficient mollicute elimination by a combination of antibiotic and antimetabolic agents.
  • Minimal toxic effects.
  • Eradicates Mycoplasma, Acholeplasma, Spiroplasma, and Entomoplasma.
  • Suited for a broad range of cell cultures.
TRUSTED_SUSTAINABILITY
Specifications
Description 1 treatment
Product Type Mycoplasma Removal
For Use With (Application) Mycoplasma elimination
Quantity 1 x 0.5 mL MycoZap™ Reagent 1, 1 x 1.5 mL MycoZap™ Reagent 2