missing translation for 'onlineSavingsMsg'
Learn More
Learn More
myo-Inositol-1,2,3,4,5,6-d6, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | myo-Inositol-1,2,3,4,5,6-d6 |
| CAS | 68922-44-1 |
| Molecular Formula | C6 2H6 H6 O6 |
| Quantity | 0.1 g |
| Synonym | myo-[1,2,3,4,5,6-2H2]Inositol, myo-Inositol-1,2,3,4,5,6-d6, myo-Inositol-C-d6, myo-Inositol-1,2,3,4,5,6-C-d6 (9 CI), Inositol-d6 (6 CI) |
| InChI Formula | InChI=1 S/C6H12O6/c7-1-2(8)4(10)6(12)5(11)3(1)9/h1-12 H/t1-,2-,3-,4+,5-,6-/i1D,2 D,3 D,4 D,5 D,6 D |
| SMILES | [2 H][C@@]1(O)[C@@]([2 H])(O)[C@@]([2 H])(O)[C@@]([2 H])(O)[C@]([2 H])(O)[C@@]1([2 H])O |
| IUPAC Name | 1,2,3,4,5,6-hexadeuteriocyclohexane-1,2,3,4,5,6-hexol |
| Molecular Weight (g/mol) | 186.19 |
| Formula Weight | 186.101 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction