missing translation for 'onlineSavingsMsg'
Learn More

myo-Inositol Hexanicotinate, TRC

Product Code. 30133022
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30133022 5 g 5g
30137915 25 g 25g
17921065 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133022 Supplier Toronto Research Chemicals Supplier No. TRCI6660705G
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6556-11-2
Molecular Formula C42 H30 N6 O12
Molecular Weight (g/mol) 810.72
Synonym Inositol Nicotinate, myo-Inositol, hexa-3-pyridinecarboxylate, Inositol, hexanicotinate, myo- (7CI,8CI), Nicotinic acid, hexaester with myo-inositol (6CI), 3-Pyridinecarboxylic acid, 1,2,3,4,5,6-cyclohexanehexayl ester, Dilcit, Dilexpal, Esantene, Hamovannid, Hexanicit, Hexanicotinoyl inositol, Hexanicotol, Hexopal, Inositol hexaniacinate, Inositol hexanicotinate, Inositol niacinate, Inositol nicotinate, Linodil, Meso-inositol hexanicotinate, Mesonex, Mesotal, Palohex, meso-Inositol hexanicotinate, myo-Inositol hexanicotinate
IUPAC Name [2,3,4,5,6-pentakis(pyridine-3-carbonyloxy)cyclohexyl] pyridine-3-carboxylate
SMILES O=C(O[C@@H]1[C@@H](OC(=O)c2cccnc2)[C@H](OC(=O)c3cccnc3)[C@@H](OC(=O)c4cccnc4)[C@@H](OC(=O)c5cccnc5)[C@H]1OC(=O)c6cccnc6)c7cccnc7

Specifications

Quantity 5 g
InChI Formula InChI=1S/C42H30N6O12/c49-37(25-7-1-13-43-19-25)55-31-32(56-38(50)26-8-2-14-44-20-26)34(58-40(52)28-10-4-16-46-22-28)36(60-42(54)30-12-6-18-48-24-30)35(59-41(53)29-11-5-17-47-23-29)33(31)57-39(51)27-9-3-15-45-21-27/h1-24,31-36H/t31-,32-,33-,34+,35-,36-
Formula Weight 810.1922
Percent Purity >95%
Chemical Name or Material Inositol Nicotinate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.