missing translation for 'onlineSavingsMsg'
Learn More

myo-Inositol Hexanicotinate, TRC

Product Code. 17921065
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17921065 1 g 1g
30133022 5 g 5g
30137915 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17921065 Supplier Toronto Research Chemicals Supplier No. TRCI6660701G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6556-11-2
InChI Formula InChI=1S/C42H30N6O12/c49-37(25-7-1-13-43-19-25)55-31-32(56-38(50)26-8-2-14-44-20-26)34(58-40(52)28-10-4-16-46-22-28)36(60-42(54)30-12-6-18-48-24-30)35(59-41(53)29-11-5-17-47-23-29)33(31)57-39(51)27-9-3-15-45-21-27/h1-24,31-36H/t31-,32-,33-,34+,35-,36-
IUPAC Name [2,3,4,5,6-pentakis(pyridine-3-carbonyloxy)cyclohexyl] pyridine-3-carboxylate
Molecular Formula C42 H30 N6 O12
Molecular Weight (g/mol) 810.72
Chemical Name or Material Inositol Nicotinate

Specifications

Quantity 1 g
Synonym Inositol Nicotinate, myo-Inositol, hexa-3-pyridinecarboxylate, Inositol, hexanicotinate, myo- (7CI,8CI), Nicotinic acid, hexaester with myo-inositol (6CI), 3-Pyridinecarboxylic acid, 1,2,3,4,5,6-cyclohexanehexayl ester, Dilcit, Dilexpal, Esantene, Hamovannid, Hexanicit, Hexanicotinoyl inositol, Hexanicotol, Hexopal, Inositol hexaniacinate, Inositol hexanicotinate, Inositol niacinate, Inositol nicotinate, Linodil, Meso-inositol hexanicotinate, Mesonex, Mesotal, Palohex, meso-Inositol hexanicotinate, myo-Inositol hexanicotinate
SMILES O=C(O[C@@H]1[C@@H](OC(=O)c2cccnc2)[C@H](OC(=O)c3cccnc3)[C@@H](OC(=O)c4cccnc4)[C@@H](OC(=O)c5cccnc5)[C@H]1OC(=O)c6cccnc6)c7cccnc7
Formula Weight 810.1922
Percent Purity >95%
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.