missing translation for 'onlineSavingsMsg'
Learn More

Myo-Inositol, Pure, Fisher Chemical

Product Code. 10723801 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
100 g
Packaging:
Plastic powder jar
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10723801 100 g Plastic powder jar 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10723801 Supplier Fisher Chemical Supplier No. I/0385/48
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 87-89-8
Melting Point 220°C
Color White
Packaging Plastic powder jar
Molecular Formula C6H12O6
MDL Number 77932
Quantity 100 g
Synonym scyllo-inositol, myo-inositol, inositol, muco-inositol, epi-inositol, i-inositol, meso-inositol, allo-inositol, 1d-chiro-inositol, myoinositol
InChI Key CDAISMWEOUEBRE-UHFFFAOYSA-N
SMILES C1(C(C(C(C(C1O)O)O)O)O)O
IUPAC Name cyclohexane-1,2,3,4,5,6-hexol
Molecular Weight (g/mol) 180.156
PubChem CID 892
ChEBI CHEBI:24848
Formula Weight 180.16g/mol
Physical Form Solid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.