missing translation for 'onlineSavingsMsg'
Learn More

myo-Inositol, TRC

Product Code. 17938024
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17938024 10 g 10g
17928324 1 g 1g
17918994 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17938024 Supplier Toronto Research Chemicals Supplier No. TRCI66599510G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 87-89-8
Molecular Formula C6 H12 O6
Molecular Weight (g/mol) 180.16
Synonym myo-Inositol, Inositol, myo- (8CI), Bios I, Cyclohexanehexol, Cyclohexitol, Dambose, Inosital, Inosite, Inositene, Inositina, Inositol, MI, Meat sugar, Mesoinosit, Mesoinosite, Mesoinositol, Mesol, Mesovit, Mouse antialopecia factor, Myoinosite, Myoinositol, Nucite, Phaseomannite, Phaseomannitol, Rat antispectacled eye factor, Scyllite, cis-1,2,3,5-trans-4,6-Cyclohexanehexol, i-Inositol, iso-Inositol, meso-Inositol
IUPAC Name cyclohexane-1,2,3,4,5,6-hexol
SMILES O[C@@H]1[C@@H](O)[C@H](O)[C@H](O)[C@H](O)[C@H]1O

Specifications

Quantity 10 g
InChI Formula InChI=1S/C6H12O6/c7-1-2(8)4(10)6(12)5(11)3(1)9/h1-12H/t1-,2-,3-,4+,5-,6-
Formula Weight 180.0634
Percent Purity >95%
Chemical Name or Material myo-Inositol
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.