Learn More
Myosin Heavy Chain Mouse anti-Multi-species, Clone: MF20, R&D Systems™
GREENER_CHOICE
Mouse Monoclonal Antibody has been used in 57 publications
Brand:  R&D Systems MAB4470-SP
Encompass
Product Code. 15796934
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
Myosin Heavy Chain Monoclonal specifically detects Myosin Heavy Chain in Multi-Species samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry.