missing translation for 'onlineSavingsMsg'
Learn More

Myosmine, 98%, may contain up to 2% water

Product Code. 11302697
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11302697 100 mg 100mg
11392687 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11302697 Supplier Thermo Scientific Alfa Aesar Supplier No. L11886.MC
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 532-12-7 | C9H10N2 | 146.193 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 532-12-7
Molecular Formula C9H10N2
Molecular Weight (g/mol) 146.193
MDL Number MFCD00052019
InChI Key DPNGWXJMIILTBS-UHFFFAOYSA-N
Synonym myosmine, 3-3,4-dihydro-2h-pyrrol-5-yl pyridine, 3-1-pyrrolin-2-yl pyridine, miosmine, unii-9o0a545w4l, 3-4,5-dihydro-3h-pyrrol-2-yl-pyridine, 2-3-pyridyl-1-pyrroline, 3-2-pyrrolin-2-yl pyridine, pyridine, 3-3,4-dihydro-2h-pyrrol-5-yl, pyridine, 3-1-pyrrolin-2-yl
PubChem CID 442649
ChEBI CHEBI:7051
IUPAC Name 3-(3,4-dihydro-2H-pyrrol-5-yl)pyridine
SMILES C1CC(=NC1)C2=CN=CC=C2

Specifications

Melting Point 42°C to 44°C
Boiling Point 82°C to 83°C (0.5 mmHg)
Quantity 100 mg
Beilstein 119254
Formula Weight 146.19
Percent Purity 98%
Assay may contain up to 2% water
Chemical Name or Material Myosmine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.