missing translation for 'onlineSavingsMsg'
Learn More

Myosmine, TRC

Product Code. 17954355
Change view
Click to view available options
Quantity:
50 mg
100 mg
250 mg
Unit Size:
100mg
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17954355 250 mg 250mg
17930995 50 mg 50mg
17994595 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17954355 Supplier Toronto Research Chemicals Supplier No. TRCM835000250MG
 Restricted Item
Estimated Shipment: 06-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 532-12-7
Molecular Formula C9 H10 N2
Molecular Weight (g/mol) 146.19
Synonym Pyridine, 3-(3,4-dihydro-2H-pyrrol-5-yl)-, Pyridine, 3-(1-pyrrolin-2-yl)- (6CI,7CI,8CI), 2-(3-Pyridyl)-1-pyrroline, Myosmine
IUPAC Name 3-(3,4-dihydro-2H-pyrrol-5-yl)pyridine
SMILES C1CN=C(C1)c2cccnc2

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C9H10N2/c1-3-8(7-10-5-1)9-4-2-6-11-9/h1,3,5,7H,2,4,6H2
Formula Weight 146.0844
Percent Purity >95%
Chemical Name or Material Myosmine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.