missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Myristic acid, MedChemExpress
Description
Myristic acid is a saturated 14-carbon fatty acid occurring in most animal and vegetable fats, particularly butterfat and coconut, palm, and nutmeg oils.
Specifications
Specifications
| Chemical Name or Material | Myristic acid |
| CAS | 544-63-8 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C14H28O2 |
| Quantity | 1 g |
| Solubility Information | DMSO : ≥ 250 mg/mL (1094.71 mM) |
| SMILES | CCCCCCCCCCCCCC(O)=O |
| Molecular Weight (g/mol) | 228.37 |
| Formula Weight | 228.37 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?