missing translation for 'onlineSavingsMsg'
Learn More

Myristicin, TRC

Product Code. 30010824
Change view
Click to view available options
Quantity:
25 mg
100 mg
250 mg
Unit Size:
100mg
250mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30010824 250 mg 250mg
30005646 100 mg 100mg
30009493 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30010824 Supplier Toronto Research Chemicals Supplier No. TRCM884600250MG
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 607-91-0
Molecular Formula C11 H12 O3
Molecular Weight (g/mol) 192.21
Synonym 1,3-Benzodioxole, 4-methoxy-6-(2-propen-1-yl)-, 1,3-Benzodioxole, 4-methoxy-6-(2-propenyl)- (9CI), Benzene, 5-allyl-1-methoxy-2,3-(methylenedioxy)- (7CI,8CI), Myristicin (6CI), 4-Methoxy-6-(2-propen-1-yl)-1,3-benzodioxole, 5-Allyl-1-methoxy-2,3-(methylenedioxy)benzene, 6-Allyl-4-methoxy-1,3-benzodioxole, Myristicine
IUPAC Name 4-methoxy-6-prop-2-enyl-1,3-benzodioxole
SMILES COc1cc(CC=C)cc2OCOc12

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C11H12O3/c1-3-4-8-5-9(12-2)11-10(6-8)13-7-14-11/h3,5-6H,1,4,7H2,2H3
Formula Weight 192.0786
Percent Purity >95%
Chemical Name or Material Myristicin
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.