missing translation for 'onlineSavingsMsg'
Learn More

Myristoleic Acid Methyl Ester, MP Biomedicals™

Product Code. 11484951 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11484951 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11484951 Supplier MP Biomedicals Supplier No. 0215576190
  • ONLINE EXCLUSIVE PRICE
    449.00 €
    425.25 € / 500mg
    Save 23.75 €
    5% Off
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Induces apoptosis and necrosis in human prostate cancer LNCaP cells. Myristoleic Acid Methyl Ester, MP Biomedicals is a cytotoxic component from the fruit extract of S. repens. It is a more hydrophobic form of the free acid.

Myristoleic acid induces apoptosis and necrosis in human prostate cancer LNCaP cells at a rate of 8.8% and 8.1%, respectively. It has a minimal inhibitory concentration (MIC) of 9μM in vivo.

Chemical Identifiers

CAS 56219-06-8
Molecular Formula C15H28O2
Molecular Weight (g/mol) 240.387
InChI Key RWIPSJUSVXDVPB-SREVYHEPSA-N
Synonym methyl myristoleate, myristoleic acid methyl ester, methyl cis-9-tetradecenoate, unii-m2a58gy964, z-methyl tetradec-9-enoate, methyl z-tetradec-9-enoate, cis-9-tetradecenoic acid, methyl ester, methyl 9z-tetradec-9-enoate, formyl 9z-tetradecenoate, methyl 9z-tetradecenoate
PubChem CID 5352674
IUPAC Name methyl (Z)-tetradec-9-enoate
SMILES CCCCC=CCCCCCCCC(=O)OC

Specifications

Refractive Index n20/D 1.446 (Lit.)
Quantity 500 mg
Formula Weight 240.4
Percent Purity ≥99%
Physical Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.