missing translation for 'onlineSavingsMsg'
Learn More

Myristoyl chloride, 97%

Product Code. 10458930 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
500g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10458930 100 g Glass bottle 100g
10635062 5 g Glass bottle 5g
10157803 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10458930 Supplier Thermo Scientific Acros Supplier No. 180631000
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 112-64-1 | C14H27ClO | 246.82 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 112-64-1
Molecular Formula C14H27ClO
Molecular Weight (g/mol) 246.82
MDL Number MFCD00000741
InChI Key LPWCRLGKYWVLHQ-UHFFFAOYSA-N
Synonym myristoyl chloride, myristoylchloride, tetradecanoylchloride, e r eao poundae, myristic acid chloride, tetradecanoic acid chloride, acmc-2099fr, ksc181q3r
PubChem CID 66986
IUPAC Name tetradecanoyl chloride
SMILES CCCCCCCCCCCCCC(=O)Cl

Tekniske data

CAS Min % 96.0
CAS Max % 100.0
Melting Point -1°C
Boiling Point 172°C to 176°C (5.0 mbar)
Flash Point >100°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)12COCl
Refractive Index 1.4480 to 1.45
Quantity 100 g
Beilstein 02, 368
Solubility Information Solubility in water: may decompose
Formula Weight 246.82
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Myristoyl chloride
Vis mere Vis mindre
compliance-icons compliance-icons
Produktidentifikator
  • Myristoyl chloride
Signalord
  • Danger
Farekategori
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Faresætning
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
  • EUH029-Contact with water liberates toxic gas.
Sikkerhedssætning
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.