missing translation for 'onlineSavingsMsg'
Learn More

Myristoyl-L-carnitine-d3 Hydrochloride, TRC

Product Code. 30477947
Change view
Click to view available options
Quantity:
2.5 mg
Unit Size:
2.5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30477947 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30477947 Supplier Toronto Research Chemicals Supplier No. TRCM8847472.5MG
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Myristoyl-L-carnitine-d3 Hydrochloride
CAS 1334532-25-0
Molecular Formula C21 D3 H39 N O4 . Cl
Quantity 2.5 mg
Synonym (R)-3-Carboxy-N,N,N-trimethyl-2-[(1-oxotetradecyl)oxy]-1-propanaminium-d3 Chloride, Tetradecanoyl-L-carnitine-(N-methyl-d3) Hydrochloride, (3R)-4-(Dimethyl-(trideuteriomethyl)azaniumyl)-3-tetradecanoyloxybutanoate hydrochloride, Tetradecanoyl-L-carnitine-d3 HCl (N-methyl-d3), Myristoyl-L-carnitine-D3 (N-methyl-D3) chloride, (2R)-3-Carboxy-N,N-dimethyl-N-(methyl-d3)-2-[(1-oxotetradecyl)oxy]-1-propanaminium chloride
InChI Formula InChI=1S/C21H41NO4.ClH/c1-5-6-7-8-9-10-11-12-13-14-15-16-21(25)26-19(17-20(23)24)18-22(2,3)4;/h19H,5-18H2,1-4H3;1H/t19-;/m1./s1/i2D3;
SMILES [Cl-].[2H]C([2H])([2H])[N+](C)(C)C[C@@H](CC(=O)O)OC(=O)CCCCCCCCCCCCC
IUPAC Name [(2R)-3-carboxy-2-tetradecanoyloxypropyl]-dimethyl-(trideuteriomethyl)azanium;chloride
Molecular Weight (g/mol) 411.034
Formula Weight 410.2991
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.