missing translation for 'onlineSavingsMsg'
Learn More

Myristyl Gallate, TRC

Product Code. 30471995
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30471995 10 g 10g
30470815 1 g 1g
30465183 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30471995 Supplier Toronto Research Chemicals Supplier No. TRCM88504310G
 Restricted Item
Estimated Shipment: 17-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18244-73-0
Molecular Formula C21H34O5
Molecular Weight (g/mol) 366.49
Synonym Gallic acid, tetradecyl ester (6CI,7CI,8CI), 1-Tetradecanol, gallate (8CI), Tetradecyl gallate, Tetradecyl 3,4,5-trihydroxybenzoate
SMILES CCCCCCCCCCCCCCOC(=O)c1cc(O)c(O)c(O)c1

Specifications

Quantity 10 g
InChI Formula InChI=1S/C21H34O5/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-26-21(25)17-15-18(22)20(24)19(23)16-17/h15-16,22-24H,2-14H2,1H3
Formula Weight 366.2406
Chemical Name or Material Myristyl Gallate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.