missing translation for 'onlineSavingsMsg'
Learn More

N-1,3,4-Thiadiazol-2-ylacetamide, TRC

Product Code. 30452510
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30452510 10 g 10g
30454673 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30452510 Supplier Toronto Research Chemicals Supplier No. TRCT34395010G
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5393-55-5
Molecular Formula C4 H5 N3 O S
Molecular Weight (g/mol) 143.17
Synonym N-(1,3,4-Thiadiazol-2-yl)acetamide, Acetazolamide Imp. B (EP)
IUPAC Name N-(1,3,4-thiadiazol-2-yl)acetamide
SMILES CC(=O)Nc1nncs1

Specifications

Quantity 10 g
InChI Formula InChI=1S/C4H5N3OS/c1-3(8)6-4-7-5-2-9-4/h2H,1H3,(H,6,7,8)
Formula Weight 143.0153
Percent Purity >95%
Chemical Name or Material N-(1,3,4-Thiadiazol-2-yl)acetamide
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.