missing translation for 'onlineSavingsMsg'
Learn More

n-[(1,3-Dimethyl-1h-pyrazol-5-yl)methyl]-n-methylamine, 97%, Thermo Scientific™

Product Code. 10654575
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10654575 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10654575 Supplier Thermo Scientific Maybridge Supplier No. CC12546DA
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
This item is not returnable. View return policy

Specifications

CAS 852227-87-3
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C7H13N3
MDL Number MFCD08700823
Quantity 1 g
Synonym n-1,3-dimethyl-1h-pyrazol-5-yl methyl-n-methylamine, 1-1,3-dimethyl-1h-pyrazol-5-yl-n-methylmethanamine, 1h-pyrazole-5-methanamine,n,1,3-trimethyl, 2,5-dimethylpyrazol-3-yl methyl methyl amine, n,1,3-trimethyl-1h-pyrazole-5-methanamine, 1,3-dimethylpyrazol-5-yl methyl methylamine, 1-2,5-dimethyl-3-pyrazolyl-n-methylmethanamine, 1-2,5-dimethylpyrazol-3-yl-n-methyl-methanamine, 1,3-dimethyl-1h-pyrazol-5-yl methyl methyl amine
InChI Key RTTFXSJNRLYKMF-UHFFFAOYSA-N
SMILES CNCC1=CC(C)=NN1C
IUPAC Name [(1,3-dimethyl-1H-pyrazol-5-yl)methyl](methyl)amine
Molecular Weight (g/mol) 139.20
PubChem CID 23005845
Formula Weight 139.2
Percent Purity 97%
Chemical Name or Material N-[(1, 3-dimethyl-1H-pyrazol-5-yl)methyl]-N-methylamine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.