missing translation for 'onlineSavingsMsg'
Learn More

N-(1-Hydroxy-2-(hydroxymethyl)-4-(4-octylphenyl)butan-2-yl)acetamide, TRC

Product Code. 30012181
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30012181 100 mg 100mg
30011753 500 mg 500mg
30004407 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30012181 Supplier Toronto Research Chemicals Supplier No. TRCH942960100MG
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 249289-10-9
Molecular Formula C21 H35 N O3
Molecular Weight (g/mol) 349.51
Synonym Acetamide, N-[1,1-bis(hydroxymethyl)-3-(4-octylphenyl)propyl]-, N-[1,1-Bis(hydroxymethyl)-3-(4-octylphenyl)propyl]acetamide, 2-[2-(4-n-Octylphenyl)ethyl]-2-acetamido-1,3-propanediol, N-(1,1-Bis(hydroxymethyl)-3-(4-octylphenyl)propyl)acetamide, N-Acetyl fingolimod
IUPAC Name N-[1-hydroxy-2-(hydroxymethyl)-4-(4-octylphenyl)butan-2-yl]acetamide
SMILES CCCCCCCCc1ccc(CCC(CO)(CO)NC(=O)C)cc1

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C21H35NO3/c1-3-4-5-6-7-8-9-19-10-12-20(13-11-19)14-15-21(16-23,17-24)22-18(2)25/h10-13,23-24H,3-9,14-17H2,1-2H3,(H,22,25)
Formula Weight 349.2617
Percent Purity >95%
Chemical Name or Material N-Acetyl Fingolimod
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.