missing translation for 'onlineSavingsMsg'
Learn More

N-(1-Naphthyl)Ethylene-Diamine Dihydrochloride, MP Biomedicals™

Product Code. 11473620 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11473620 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11473620 Supplier MP Biomedicals Supplier No. 0210239705
 Restricted Item
Estimated Shipment: 12-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

N-(1-Naphthyl)Ethylene-Diamine Dihydrochloride, MP Biomedicals is used for colorimetric determination of sulfonamide in blood.

Specifications

CAS 1465-25-4
Melting Point 188°C to 198°C
Molecular Formula C12H16Cl2N2
Quantity 5 g
Synonym n-1-naphthyl ethylenediamine dihydrochloride, n1-naphthalen-1-yl ethane-1,2-diamine dihydrochloride, marshall's reagent, n-1-naphthylethylenediamine dihydrochloride, ccris 425, unii-h734599kjl, 1,2-ethanediamine, n-1-naphthalenyl-, dihydrochloride, 2-1-naphthylamino ethylamine 2hcl, bratton-marshall reagent, n-1-naphthalenyl-1,2-ethanediamine dihydrochloride
Solubility Information Soluble in water (20mg/mL; almost clear, colorless to slightly yellow solution) and 25mg/mL, EtOH.
InChI Key MZNYWPRCVDMOJG-UHFFFAOYSA-N
SMILES C1=CC=C2C(=C1)C=CC=C2NCCN.Cl.Cl
IUPAC Name N'-naphthalen-1-ylethane-1,2-diamine;dihydrochloride
Molecular Weight (g/mol) 259.174
PubChem CID 15106
ChEBI CHEBI:53452
Formula Weight 259.18
Physical Form Powder
Show More Show Less
compliance-icons
Product Identifier
  • N-2-aminoéthyl-1-naphtylamine, dichlorhydrate
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501-Dispose of contents/container to .
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.