missing translation for 'onlineSavingsMsg'
Learn More

N-(1-Naphthyl)ethylenediamine, dihydrochloride, 98+%, ACS reagent

Product Code. 10219370 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass Bottle
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 10219370

Brand: Thermo Scientific Acros 423991000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

N-(1-Naphthyl)ethylenediamine, dihydrochloride, 98+%, C12H14N2.2ClH, CAS Number-1465-25-4 | CAS: 1465-25-4 | C12H16Cl2N2 | 259.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1465-25-4
MDL Number MFCD00012556
InChI Key MZNYWPRCVDMOJG-UHFFFAOYSA-N
Synonym n-1-naphthyl ethylenediamine dihydrochloride, n1-naphthalen-1-yl ethane-1,2-diamine dihydrochloride, marshall's reagent, n-1-naphthylethylenediamine dihydrochloride, ccris 425, unii-h734599kjl, 1,2-ethanediamine, n-1-naphthalenyl-, dihydrochloride, 2-1-naphthylamino ethylamine 2hcl, bratton-marshall reagent, n-1-naphthalenyl-1,2-ethanediamine dihydrochloride
PubChem CID 15106
ChEBI CHEBI:53452

Specifications

CAS Min % 98.0
CAS Max % 100.0
Decomposition Information 200°C
Color Beige to White or Pink
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Glass Bottle
Molecular Formula C12H16Cl2N2
Linear Formula C10H7NHCH2CH2NH2·2HCl
Quantity 100 g
Beilstein 12, II, 699
Sensitivity (to sulfanilamide) Passes
Merck Index 15, 6491
Solubility Information Solubility in water: soluble. Other solubilities: soluble in hot water, slightly soluble in acetone and absolute alcohol
SMILES C1=CC=C2C(=C1)C=CC=C2NCCN.Cl.Cl
IUPAC Name N'-naphthalen-1-ylethane-1,2-diamine;dihydrochloride
Molecular Weight (g/mol) 259.174
Formula Weight 259.18
Percent Purity ≥98.0% (on dry substance)
Grade ACS Reagent
Physical Form Powder
Chemical Name or Material N-(1-Naphthyl)ethylenediamine, dihydrochloride
Show More Show Less
compliance-icons
Product Identifier
  • N-(1-Naphthyl)ethylenediamine, dihydrochloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.