missing translation for 'onlineSavingsMsg'
Learn More

N-(1-Naphthyl)ethylenediamine dihydrochloride, ACS reagent, ≥98%, Honeywell Fluka™

Product Code. p-7236095 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16675823 5 g 5g
16665823 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16675823 Supplier Honeywell Fluka Supplier No. 33461H5G
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 1465-25-4
Molecular Formula C12H16Cl2N2
Molecular Weight (g/mol) 259.17
InChI Key MZNYWPRCVDMOJG-UHFFFAOYSA-N
IUPAC Name dihydrogen N1-(naphthalen-1-yl)ethane-1,2-diamine dichloride
SMILES [H+].[H+].[Cl-].[Cl-].NCCNC1=C2C=CC=CC2=CC=C1

Specifications

CAS Max % 1
Melting Point 194°C to 198°C (Decomposition)
Color Colorless
Quantity 5 g
Chemical Name or Material N-(1-Naphthyl)ethylenediamine dihydrochloride
compliance-icons
Product Identifier
  • N-(1-Naphthyl)-Ethylendiamine dihydrochloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: N-2-Aminoethyl-1-naphthylamine dihydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.