missing translation for 'onlineSavingsMsg'
Learn More

N-(1-Naphthyl)ethylenediamine dihydrochloride, Honeywell Fluka™

Product Code. 16665763 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16665763 25 g 25g
16655763 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16665763 Supplier Honeywell Research Chemicals Supplier No. 222488H25G
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

ACS reagent, >98%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1465-25-4
Molecular Formula C12H16Cl2N2
Molecular Weight (g/mol) 259.17
InChI Key MZNYWPRCVDMOJG-UHFFFAOYSA-N
PubChem CID 15106
ChEBI CHEBI:53452
IUPAC Name dihydrogen N1-(naphthalen-1-yl)ethane-1,2-diamine dichloride
SMILES [H+].[H+].[Cl-].[Cl-].NCCNC1=C2C=CC=CC2=CC=C1

Specifications

Packaging Plastic Bottle
Quantity 25 g
Percent Purity min. 98 %
Physical Form Solid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.