missing translation for 'onlineSavingsMsg'
Learn More

N-1-Naphthylethylenediamine Dihydrochloride, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™

Product Code. 15185083 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
25 g
Packaging:
Wide Mouth Amber Glass Bottle
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15185083 25 g Wide Mouth Amber Glass Bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15185083 Supplier Avantor J.T.Baker Supplier No. R70103

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High quality chemicals for laboratory and specialized industrial use

    For Nitrogen Dioxide Determination (ASTM D-1607)

Chemical Identifiers

CAS 1465-25-4
Molecular Formula C12H16Cl2N2
Molecular Weight (g/mol) 259.174
InChI Key MZNYWPRCVDMOJG-UHFFFAOYSA-N
Synonym n-1-naphthyl ethylenediamine dihydrochloride, n1-naphthalen-1-yl ethane-1,2-diamine dihydrochloride, marshall's reagent, n-1-naphthylethylenediamine dihydrochloride, ccris 425, unii-h734599kjl, 1,2-ethanediamine, n-1-naphthalenyl-, dihydrochloride, 2-1-naphthylamino ethylamine 2hcl, bratton-marshall reagent, n-1-naphthalenyl-1,2-ethanediamine dihydrochloride
PubChem CID 15106
ChEBI CHEBI:53452
IUPAC Name N'-naphthalen-1-ylethane-1,2-diamine;dihydrochloride
SMILES C1=CC=C2C(=C1)C=CC=C2NCCN.Cl.Cl

Specifications

Packaging Wide Mouth Amber Glass Bottle
Quantity 25 g
Formula Weight 259.18
Grade BAKER ANALYZED™ ACS Reagent
Chemical Name or Material N-1-Naphthylethylenediamine Dihydrochloride
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.