missing translation for 'onlineSavingsMsg'
Learn More

N-(1-Naphthylmethyl)methylamine Hydrochloride, TRC

Product Code. 30137965
Change view
Click to view available options
Quantity:
1 g
25 g
Unit Size:
1g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137965 25 g 25g
17972315 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30137965 Supplier Toronto Research Chemicals Supplier No. TRCN37295025G
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 65473-13-4
Molecular Formula C12 H13 N . Cl H
Molecular Weight (g/mol) 207.7
Synonym N-Methyl-C-(naphthalen-1-yl)methanamine Hydrochloride, Terbinafine Hydrochloride Imp. A (EP) as Hydrochloride, Secondary Amine Hydrochloride of Terbinafine
IUPAC Name N-methyl-1-naphthalen-1-ylmethanamine;hydrochloride
SMILES Cl.CNCc1cccc2ccccc12

Specifications

Quantity 25 g
InChI Formula InChI=1S/C12H13N.ClH/c1-13-9-11-7-4-6-10-5-2-3-8-12(10)11;/h2-8,13H,9H2,1H3;1H
Formula Weight 207.0815
Chemical Name or Material N-Methyl-C-(naphthalen-1-yl)methanamine Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.