missing translation for 'onlineSavingsMsg'
Learn More

N-(1-Naphthylmethyl)methylamine Hydrochloride, TRC

Product Code. 17972315
Change view
Click to view available options
Quantity:
1 g
25 g
Unit Size:
1g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17972315 1 g 1g
30137965 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17972315 Supplier Toronto Research Chemicals Supplier No. TRCN3729501G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 65473-13-4
Formula Weight 207.0815
InChI Formula InChI=1S/C12H13N.ClH/c1-13-9-11-7-4-6-10-5-2-3-8-12(10)11;/h2-8,13H,9H2,1H3;1H
IUPAC Name N-methyl-1-naphthalen-1-ylmethanamine;hydrochloride
Molecular Formula C12 H13 N . Cl H
Molecular Weight (g/mol) 207.7
SMILES Cl.CNCc1cccc2ccccc12
Synonym N-Methyl-C-(naphthalen-1-yl)methanamine Hydrochloride, Terbinafine Hydrochloride Imp. A (EP) as Hydrochloride, Secondary Amine Hydrochloride of Terbinafine
Chemical Name or Material N-Methyl-C-(naphthalen-1-yl)methanamine Hydrochloride

Specifications

Quantity 1 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.