missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Aluminum Crimp Closures with Center Hole, N11

Product Code. 10326701 Shop All Macherey-Nagel Products
Click to view available options
Color:
Silver
Unit Size:
Pack of 100
This item is not returnable. View return policy

Product Code. 10326701

Brand: Macherey-Nagel™ 70256

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Enables needle penetration with reduced handling and contamination risk. Macherey-Nagel™ Aluminum Crimp Closures with Center Hole, N11 are pre-assembled, ready to use crimp caps with medium-soft septa for use with N11 crimp neck chromatography vials.

Fully automated closure manufacturing with defined production parameters avoids variations in the physical and analytical condition of the cap. Strict analytical specifications allow only processing of septa material that passes the comprehensive analytical tests in the lab. These products are manufactured under strict hygienic conditions. In-process controls and a final QC inspection have been implemented to grant reliable product quality.

  • Pre-assembled, ready-to-use crimp closures are an economical option
  • Used with Macherey-Nagel™ N11 crimp neck chromatography vials  featuring an 11mm opening
  • Made of sturdy silver aluminum with septum in the center hole
  • Septum is made of natural rubber, red-orange butyl and a colorless TEF membrane
  • 100 caps are packaged in a polyethylene (PE) bag

Specifications

Color Silver
Material Aluminum, Butyl Rubber, Rubber, TEF
Septum Natural rubber / Butyl red orange/ TEF colorless
For Use With (Application) Chromatography
Includes Center hole and Septum
Quantity 100 Pk.
Thickness (Metric) 1 mm
Type Crimp Cap
Closure Size N11
For Use With (Equipment) N 11 Crimp Neck Vials
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.