missing translation for 'onlineSavingsMsg'
Learn More

N-[(1S)-1-(Hydroxymethyl)-2-[(3S)-2-oxo-3-pyrrolidinyl]ethyl]-carbamic Acid tert-Butyl Ester, TRC

Product Code. 30407436
Change view
Click to view available options
Quantity:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30407436 25 mg
30399715 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30407436 Supplier Toronto Research Chemicals Supplier No. TRCH94778525MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 249736-45-6
Molecular Formula C12H22N2O4
Molecular Weight (g/mol) 258.31
Synonym N-[(1S)-1-(Hydroxymethyl)-2-[(3S)-2-oxo-3-pyrrolidinyl]ethyl]-carbamic Acid 1,1-Dimethylethyl Ester
IUPAC Name tert-butyl N-[(2S)-1-hydroxy-3-[(3S)-2-oxopyrrolidin-3-yl]propan-2-yl]carbamate
SMILES CC(C)(C)OC(=O)N[C@H](CO)C[C@@H]1CCNC1=O

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C12H22N2O4/c1-12(2,3)18-11(17)14-9(7-15)6-8-4-5-13-10(8)16/h8-9,15H,4-7H2,1-3H3,(H,13,16)(H,14,17)/t8-,9-/m0/s1
Formula Weight 258.158
Chemical Name or Material N-[(1S)-1-(Hydroxymethyl)-2-[(3S)-2-oxo-3-pyrrolidinyl]ethyl]-carbamic Acid tert-Butyl Ester
Product Title
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.