missing translation for 'onlineSavingsMsg'
Learn More

N-[2-[2-(2-Chloro-4,6-dinitrophenyl)diazenyl]-5-(diethylamino)phenyl]acetamide (>90%), TRC

Product Code. 30358640
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30358640 50 mg -
30350322 10 mg 10mg
30352424 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30358640 Supplier Toronto Research Chemicals Supplier No. TRCC54613050MG
 Restricted Item
Estimated Shipment: 01-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 66557-45-7
Molecular Formula C18H19ClN6O5
Molecular Weight (g/mol) 434.83
Synonym Acetamide, N-[2-[2-(2-chloro-4,6-dinitrophenyl)diazenyl]-5-(diethylamino)phenyl]-, Acetamide, N-[2-[(2-chloro-4,6-dinitrophenyl)azo]-5-(diethylamino)phenyl]- (9CI), Disperse Violet 93 (Cl derivative)
IUPAC Name N-[2-[(E)-(2-chloro-4,6-dinitrophenyl)diazenyl]-5-(diethylamino)phenyl]acetamide
SMILES CCN(CC)c1ccc(N=Nc2c(Cl)cc(cc2[N+](=O)[O-])[N+](=O)[O-])c(NC(=O)C)c1

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C18H19ClN6O5/c1-4-23(5-2)12-6-7-15(16(9-12)20-11(3)26)21-22-18-14(19)8-13(24(27)28)10-17(18)25(29)30/h6-10H,4-5H2,1-3H3,(H,20,26)
Formula Weight 434.1105
Chemical Name or Material Disperse Violet 93 (Cl derivative)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.