missing translation for 'onlineSavingsMsg'
Learn More

N-{[2-(3-Chlorophenyl)-1,3-thiazol-4-yl]methyl}-N-methylamine, 90%, Thermo Scientific™

Product Code. 10183084
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10183084 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10183084 Supplier Thermo Scientific Maybridge Supplier No. CC46746DA
 Restricted Item
Estimated Shipment: 19-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 864068-99-5
CAS Min % 89.5
CAS Max % 100.0
Assay Percent Range 90%
Packaging Amber Glass Bottle
Molecular Formula C11H11ClN2S
MDL Number MFCD08690268
Quantity 1 g
Synonym 4-thiazolemethanamine,2-3-chlorophenyl-n-methyl, 2-3-chlorophenyl-4-methylamino methyl-1,3-thiazole, 2-3-chlorophenyl-1,3-thiazol-4-yl methyl methyl amine, 1-2-3-chlorophenyl thiazol-4-yl-n-methylmethanamine, 2-3-chlorophenyl 1,3-thiazol-4-yl methyl methylamine, 1-2-3-chlorophenyl-1,3-thiazol-4-yl-n-methylmethanamine, 1-2-3-chlorophenyl-1,3-thiazol-4-yl-n-methylmethylamine, n-2-3-chlorophenyl-1,3-thiazol-4-yl methyl-n-methylamine
InChI Key YHQACXKNKUPSSA-UHFFFAOYSA-N
SMILES CNCC1=CSC(=N1)C2=CC(=CC=C2)Cl
IUPAC Name 1-[2-(3-chlorophenyl)-1,3-thiazol-4-yl]-N-methylmethanamine
Molecular Weight (g/mol) 238.733
PubChem CID 18525799
Formula Weight 238.74
Percent Purity 90%
Chemical Name or Material N-{[2-(3-chlorophenyl)-1, 3-thiazol-4-yl]methyl}-N-methylamine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.