missing translation for 'onlineSavingsMsg'
Learn More

N-(2,4-Dichlorophenyl)maleamic acid, 97%, Thermo Scientific™

Product Code. 11431467
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11431467 25 g 25g
11411467 1 g 1g
11421467 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11431467 Supplier Thermo Scientific Alfa Aesar Supplier No. B24941.14
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95695-47-9
Molecular Formula C10H7Cl2NO3
Molecular Weight (g/mol) 260.07
MDL Number MFCD00082905
InChI Key WFWKZRKAECLVSE-ARJAWSKDSA-N
Synonym n-2,4-dichlorophenyl maleamic acid, 2z-3-2,4-dichlorophenyl carbamoyl prop-2-enoic acid, 4-2,4-dichloroanilino-4-oxobut-2-enoic acid, n-2,4-dichlorophenyl maleamidic acid, z-4-2,4-dichloroanilino-4-oxobut-2-enoic acid, z-4-2,4-dichlorophenyl amino-4-oxobut-2-enoic acid
PubChem CID 1549984
IUPAC Name (Z)-4-(2,4-dichloroanilino)-4-oxobut-2-enoic acid
SMILES C1=CC(=C(C=C1Cl)Cl)NC(=O)C=CC(=O)O

Specifications

Melting Point 177°C to 180°C
Quantity 25 g
Formula Weight 260.07
Percent Purity 97%
Chemical Name or Material N-(2,4-Dichlorophenyl)maleamic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.