missing translation for 'onlineSavingsMsg'
Learn More

N-(2,5-Dichlorophenyl)maleamic acid, 97%, Thermo Scientific™

Product Code. 11497016
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11497016 5 g 5g
11407026 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11497016 Supplier Thermo Scientific Alfa Aesar Supplier No. B25342.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 31460-32-9
Molecular Formula C10H7Cl2NO3
Molecular Weight (g/mol) 260.07
MDL Number MFCD00082906
InChI Key WSKTYRGKGHEEDM-ARJAWSKDSA-N
Synonym z-4-2,5-dichlorophenyl amino-4-oxobut-2-enoic acid, n-2,5-dichlorophenyl maleamic acid, 2z-3-2,5-dichlorophenyl carbamoyl prop-2-enoic acid, 2z-3-n-2,5-dichlorophenyl carbamoyl prop-2-enoic acid, 4-2,5-dichloroanilino-4-oxobut-2-enoic acid, n-2,5-dichlorophenyl maleamidic acid, 4-2,5-dichloroanilino-4-oxo-2-butenoic acid, z-4-2,5-dichloroanilino-4-oxobut-2-enoic acid
PubChem CID 1551520
IUPAC Name (Z)-4-(2,5-dichloroanilino)-4-oxobut-2-enoic acid
SMILES C1=CC(=C(C=C1Cl)NC(=O)C=CC(=O)O)Cl

Specifications

Melting Point 140°C to 144°C
Quantity 5 g
Formula Weight 260.07
Percent Purity 97%
Chemical Name or Material N-(2,5-Dichlorophenyl)maleamic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.