missing translation for 'onlineSavingsMsg'
Learn More

N-(2-Bromoethyl-d4)phthalimide, CDN

Product Code. 30322216
Change view
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30322216 0.5 g 0.5g
30319624 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30322216 Supplier CDN Supplier No. CDND78580.5G
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.

TRUSTED_SUSTAINABILITY

Specifikationer

Chemical Name or Material N-(2-Bromoethyl-d4)phthalimide
CAS 1009307-39-4
Molecular Formula C10 D4 H4 Br N O2
Quantity 0.5 g
Synonym 1 H-Isoindole-1,3(2 H)-dione, 2-(2-bromoethyl-1,1,2,2-d4)- (ACI), 2-(2-Bromoethyl-1,1,2,2-d4)-1 H-isoindole-1,3(2 H)-dione (ACI), 2-(2-Bromo-1,1,2,2-tetradeuterioethyl)isoindole-1,3-dione, 2-(2-Bromoethyl-d4)phthalimide, N-(2-Bromoethyl-d4)phthalimide, N-(2-Bromoethyl)phthalimide-d4, N-(2-Bromoethyl)phthalimide-D4 (2-bromoethyl-D4)
InChI Formula InChI=1 S/C10H8BrNO2/c11-5-6-12-9(13)7-3-1-2-4-8(7)10(12)14/h1-4 H,5-6H2/i5D2,6D2
SMILES [2 H]C([2 H])(Br)C([2 H])([2 H])N1C(=O)c2ccccc2C1=O
IUPAC Name 2-(2-bromo-1,1,2,2-tetradeuterioethyl)isoindole-1,3-dione
Molecular Weight (g/mol) 258.105
Formula Weight 256.999 g/mol
Percent Purity 99 atom % D, min 98% Chemical Purity
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.