missing translation for 'onlineSavingsMsg'
Learn More

N-(2-Bromoethyl)phthalimide, TRC

Product Code. 30329607
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30329607 25 g 25g
30346124 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30329607 Supplier Toronto Research Chemicals Supplier No. TRCB68397525G
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-(2-Bromoethyl)phthalimide
CAS 574-98-1
Molecular Formula C10H8BrNO2
Quantity 25 g
Synonym 2-(2-Bromoethyl)isoindoline-1,3-dione, 1-Bromo-2-phthalimidoethane, 2-(1,3-Dioxo-1,3-dihydro-isoindol-2-yl)-ethyl Bromide, 2-(2-Bromoethyl)-1H-isoindole-1,3(2H)-dione, 2-(2-Bromoethyl)-2,3-dihydro-1H-isoindole-1,3-dione, 2-(2-Bromoethyl)isoindole-1,3-dione, 2-(2-Bromoethyl)isoindoline-1,3-dione, 2-(2-Bromoethyl)phthalimide, 2-Phthalimidoethyl Bromide, N-(2-Bromoethyl)phthalimide, N-(beta-Bromoethyl)phthalimide, NSC 2688, beta-Phthalimidoethyl Bromide
InChI Formula InChI=1S/C10H8BrNO2/c11-5-6-12-9(13)7-3-1-2-4-8(7)10(12)14/h1-4H,5-6H2
SMILES BrCCN1C(=O)c2ccccc2C1=O
IUPAC Name 2-(2-bromoethyl)isoindole-1,3-dione
Molecular Weight (g/mol) 254.08
Formula Weight 252.9738
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.