missing translation for 'onlineSavingsMsg'
Learn More

N-(2-Carboxyethyl)iminodiacetic Acid, ≥98.0%, Honeywell™ Fluka™

Product Code. 15618480 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
5 g
Unit Size:
5g
This item is not returnable. View return policy

Product Code. 15618480

Brand: Honeywell-Fluka 925775G

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

for complexometry,≥98.0%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Specifications

CAS 6245-75-6
Melting Point 195°C
Color White
Molecular Formula C7H11NO6
MDL Number MFCD00059634
Quantity 5 g
UN Number NONH for all modes of transport
Beilstein 1712227
Synonym n-2-carboxyethyl iminodiacetic acid, beta-alanine diacetic acid, unii-kou355f9z2, n,n-bis carboxymethyl-, a-alanine, .beta.-alanine, n,n-bis carboxymethyl, 3-bis carboxymethyl amino propanoic acid, n-2-carboxyethyl dimethylsulfonium chloride, n,n-bis carboxymethyl-beta-alanine, beta-alaninediacetic acid, nitrilodiacetic-propionic acid
InChI Key UWRBFYBQPCJRRL-UHFFFAOYSA-N
SMILES OC(=O)CCN(CC(O)=O)CC(O)=O
IUPAC Name 3-[bis(carboxymethyl)amino]propanoic acid
Molecular Weight (g/mol) 205.17
PubChem CID 535795
Formula Weight 205.17g/mol
Percent Purity ≥98.0%
Chemical Name or Material N-(2-Carboxyethyl)iminodiacetic acid, For complexometry
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.