missing translation for 'onlineSavingsMsg'
Learn More

N-(2-Chloro-1-oxopropyl)glycine, TRC

Product Code. 30011238
Change view
Click to view available options
Quantity:
250 mg
1 g
2.5 g
Unit Size:
1g
2.5g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30011238 2.5 g 2.5g
30133360 1 g 1g
30137886 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30011238 Supplier Toronto Research Chemicals Supplier No. TRCC5961502.5G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 85038-45-5
Molecular Formula C5H8ClNO3
Molecular Weight (g/mol) 165.57
Synonym N-(2-Chloropropionyl)glycine, Tiopronin Impuriy 1
IUPAC Name 2-(2-chloropropanoylamino)acetic acid
SMILES CC(Cl)C(=O)NCC(=O)O

Specifications

InChI Formula InChI=1S/C5H8ClNO3/c1-3(6)5(10)7-2-4(8)9/h3H,2H2,1H3,(H,7,10)(H,8,9)
Quantity 2.5 g
Formula Weight 165.0193
Shipping Condition Room Temperature
Chemical Name or Material N-(2-Chloro-1-oxopropyl)glycine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.