missing translation for 'onlineSavingsMsg'
Learn More

N-[2-Chloro-3-(dimethylamino)-2-propen-1-ylidene]-N-methylmethanaminium Hexafluorophosphate, TRC

Product Code. 30368537
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30368537 1 g 1g
30355535 10 g 10g
30350508 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30368537 Supplier Toronto Research Chemicals Supplier No. TRCC2933251G
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 249561-98-6
Molecular Formula C7 H14 Cl N2 . F6 P
Molecular Weight (g/mol) 306.62
Synonym Methanaminium, N-[2-chloro-3-(dimethylamino)-2-propen-1-ylidene]-N-methyl-, hexafluorophosphate(1-) (1:1), Methanaminium, N-[2-chloro-3-(dimethylamino)-2-propenylidene]-N-methyl-, hexafluorophosphate(1-) (9CI)
IUPAC Name [2-chloro-3-(dimethylamino)prop-2-enylidene]-dimethylazanium;hexafluorophosphate
SMILES CN(C)C=C(Cl)C=[N+](C)C.F[P-](F)(F)(F)(F)F

Specifications

Quantity 1 g
InChI Formula InChI=1S/C7H14ClN2.F6P/c1-9(2)5-7(8)6-10(3)4;1-7(2,3,4,5)6/h5-6H,1-4H3;/q+1;-1
Formula Weight 306.0487
Chemical Name or Material N-[2-Chloro-3-(dimethylamino)-2-propenylidene]-N-methyl-methanaminium Hexafluorophosphate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.