missing translation for 'onlineSavingsMsg'
Learn More

N-(2-Cyanoethyl)-N-(2-hydroxyethyl)aniline, 98%, Thermo Scientific™

Product Code. 11327626
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11327626 50 g 50g
11317626 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11327626 Supplier Thermo Scientific Alfa Aesar Supplier No. L11183.18

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
N-(2-Cyanoethyl)-N-(2-hydroxyethyl)aniline is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Solubility
Not miscible or difficult to mix with water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with bases and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 92-64-8
Molecular Formula C11H14N2O
Molecular Weight (g/mol) 190.246
MDL Number MFCD00035705
InChI Key WZJJWQVBLSPALW-UHFFFAOYSA-N
Synonym n-cyanoethyl-n-hydroxyethylaniline, n-2-cyanoethyl-n-2-hydroxyethyl aniline, emery 5724, n-cyanoethyl-hydroxyethyl aniline, propanenitrile, 3-2-hydroxyethyl phenylamino, n-cyanoethyl-n-hydroxyethyl aniline, aniline, n-cyanoethyl-n-hydroxyethyl, 3-2-hydroxyethyl phenylamino propanenitrile, 3-n-2-hydroxyethyl anilino propiononitrile, propionitrile, 3-2-hydroxyethyl phenylamino
PubChem CID 113349
IUPAC Name 3-[N-(2-hydroxyethyl)anilino]propanenitrile
SMILES C1=CC=C(C=C1)N(CCC#N)CCO

Specifications

Density 1.12
Boiling Point 107°C to 110°C (15 mmHg)
Refractive Index 1.5705
Quantity 50 g
Beilstein 2369849
Solubility Information Not miscible or difficult to mix with water.
Formula Weight 190.25
Percent Purity 98%
Chemical Name or Material N-(2-Cyanoethyl)-N-(2-hydroxyethyl)aniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.