missing translation for 'onlineSavingsMsg'
Learn More

N,2-Dimethyl-(1,3-thiazol-4-yl)methylamine, 97%, Thermo Scientific™

Product Code. 11380557
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11380557 250 mg Amber Glass Bottle 250mg
10682845 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11380557 Supplier Thermo Scientific Maybridge Supplier No. CC05746CB
 Restricted Item
Estimated Shipment: 23-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 144163-81-5
Molecular Formula C6H10N2S
Molecular Weight (g/mol) 142.22
MDL Number MFCD06738964
InChI Key GJHOBSIOQKGKSG-UHFFFAOYSA-N
Synonym methyl-2-methyl-thiazol-4-ylmethyl-amine, methyl-2-methylthiazole-4-yl methyl amine, n-methyl-1-2-methylthiazol-4-yl methanamine, methyl 2-methyl-1,3-thiazol-4-yl methyl amine, 4-thiazolemethanamine,n,2-dimethyl, n-methyl 2-methylthiazol-4-yl methanamine, 4-thiazolemethanamine, n,2-dimethyl, n-methyl-1-2-methyl-1,3-thiazol-4-yl methanamine, n,2-dimethyl-1,3-thiazol-4-yl methylamine, 2-methyl-4-methyl aminomethyl-1,3-thiazole
PubChem CID 18939489
IUPAC Name N-methyl-1-(2-methyl-1,3-thiazol-4-yl)methanamine
SMILES CC1=NC(=CS1)CNC

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 142.23
Percent Purity 97%
Chemical Name or Material N, 2-dimethyl-(1, 3-thiazol-4-yl)methylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.