missing translation for 'onlineSavingsMsg'
Learn More

N-(2-Hydroxyethyl)hexamethyleneimine, 95%

Product Code. 11392536
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11392536 5 g 5g
11302546 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11392536 Supplier Thermo Scientific Alfa Aesar Supplier No. L06497.06
 Restricted Item
Estimated Shipment: 30-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 20603-00-3 | C8H17NO | 143.23 g/mol

N-(2-Hydroxyethyl)hexamethyleneimine is used in organic synthesis, building blocks. It is mainly involved as an intermediate in the synthesis of Bazedoxifene acetate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-(2-Hydroxyethyl)hexamethyleneimine is used in organic synthesis, building blocks. It is mainly involved as an intermediate in the synthesis of Bazedoxifene acetate.

Solubility
Fully miscible in water.

Notes
Store away from air and oxidizing agents. Store under dry inert gas. Incompatible with air and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20603-00-3
Molecular Formula C8H17NO
Molecular Weight (g/mol) 143.23
MDL Number MFCD00020988
InChI Key VMRYMOMQCYSPHS-UHFFFAOYSA-N
Synonym 2-azepan-1-yl ethanol, hexahydro-1h-azepine-1-ethanol, n-2-hydroxyethyl hexamethyleneimine, 2-azepan-1-yl ethan-1-ol, 2-hexamethyleneimino ethanol, 2-azaperhydroepinylethan-1-ol, 2-azepan-1-ylethan-1-ol, 2-azepan-1-yl ;ethanol, 2-n-hexamethyleneimino ethanol, 1-2-hydroxyethyl homopiperidine
PubChem CID 88615
IUPAC Name 2-(azepan-1-yl)ethanol
SMILES C1CCCN(CC1)CCO

Specifications

Density 0.971
Boiling Point 114°C to 115°C (23 mmHg)
Refractive Index 1.484
Quantity 5 g
UN Number UN2735
Beilstein 104110
Sensitivity Air Sensitive
Solubility Information Fully miscible in water.
Formula Weight 143.22
Percent Purity 95%
Chemical Name or Material N-(2-Hydroxyethyl)hexamethyleneimine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.