missing translation for 'onlineSavingsMsg'
Learn More

N-(2-Hydroxyethyl)morpholine, 99%

Product Code. 10041070 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
500g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10041070 100 g Glass bottle 100g
10598470 5 g Glass bottle 5g
10605742 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Product Code. 10041070 Supplier Thermo Scientific Acros Supplier No. 121181000
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

CAS: 622-40-2 | C6H13NO2 | 131.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 622-40-2
Molecular Formula C6H13NO2
Molecular Weight (g/mol) 131.17
MDL Number MFCD00006180
InChI Key KKFDCBRMNNSAAW-UHFFFAOYSA-N
Synonym 2-morpholinoethanol, 4-2-hydroxyethyl morpholine, 4-morpholineethanol, n-2-hydroxyethyl morpholine, 2-4-morpholinyl ethanol, morpholine ethanol, morpholineethanol, hydroxyethylmorpholine, ethanol, 2-morpholino, beta-morpholinoethanol
PubChem CID 61163
ChEBI CHEBI:67144
IUPAC Name 2-morpholin-4-ylethanol
SMILES C1COCCN1CCO

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -1.0°C
Color Colorless to Yellow
Density 1.0800g/mL
Boiling Point 223.0°C to 225.0°C
Flash Point 107°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Refractive Index 1.4760 to 1.4780
Quantity 100 g
Beilstein 27, 7
Specific Gravity 1.08
Solubility Information Solubility in water: miscible
Viscosity 26.6 mPa.s (20°C)
Formula Weight 131.17
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material N-(2-Hydroxyethyl)morpholine, 99%
Show More Show Less
compliance-icons
Product Identifier
  • N-(2-Hydroxyethyl)morpholine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.