missing translation for 'onlineSavingsMsg'
Learn More

N-(2-Hydroxyethyl)phthalimide, 99%

Product Code. 10593091 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10593091 100 g Glass bottle 100g
10583091 25 g Glass bottle 25g
10455485 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10593091 Supplier Thermo Scientific Acros Supplier No. 124921000
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 3891-07-4 | C10H9NO3 | 191.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3891-07-4
Molecular Formula C10H9NO3
Molecular Weight (g/mol) 191.19
MDL Number MFCD00005903
InChI Key MWFLUYFYHANMCM-UHFFFAOYSA-N
Synonym 2-1,2,2-trimethylpropylcarbamoyl benzoic acid, 2-3,3-dimethylbutan-2-yl carbamoyl benzoic acid, 2-1,2,2-trimethylpropyl amino carbonyl benzoic acid, n-pinacolylphthalamic acid, 2-3,3-dimethylbutan-2-ylcarbamoyl benzoic acid, +-n-pinacolylphthalamic acid
PubChem CID 3354762
IUPAC Name 2-(3,3-dimethylbutan-2-ylcarbamoyl)benzoic acid
SMILES OCCN1C(=O)C2=CC=CC=C2C1=O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 126°C to 129°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (on dry substance) (HPLC)
Packaging Glass bottle
Quantity 100 g
Formula Weight 191.19
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material N-(2-Hydroxyethyl)phthalimide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.