missing translation for 'onlineSavingsMsg'
Learn More

N-(2-Hydroxyethyl)piperazine(1-Piperazineethanol), TRC

Product Code. 30420723
Change view
Click to view available options
Quantity:
1 g
10 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30420723 1 g
30413296 100 g
30413770 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30420723 Supplier Toronto Research Chemicals Supplier No. TRCP4801201G
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103-76-4
Molecular Formula C6 H14 N2 O
Molecular Weight (g/mol) 130.19
Synonym 1-(2-Hydroxyethyl)piperazine
IUPAC Name 2-piperazin-1-ylethanol
SMILES OCCN1CCNCC1

Specifications

Quantity 1 g
InChI Formula InChI=1S/C6H14N2O/c9-6-5-8-3-1-7-2-4-8/h7,9H,1-6H2
Formula Weight 130.1106
Chemical Name or Material 1-(2-Hydroxyethyl)piperazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.