missing translation for 'onlineSavingsMsg'
Learn More

N-(2-Hydroxyethyl)piperazine, 98.5%

Product Code. 10704061 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10704061 100 g Glass bottle 100g
10498840 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10704061 Supplier Thermo Scientific Acros Supplier No. 121191000
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 103-76-4 | C6H14N2O | 130.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103-76-4
Molecular Formula C6H14N2O
Molecular Weight (g/mol) 130.19
MDL Number MFCD00005970
InChI Key WFCSWCVEJLETKA-UHFFFAOYSA-N
Synonym n-2-hydroxyethyl piperazine, 1-piperazineethanol, 1-2-hydroxyethyl piperazine, hydroxyethylpiperazine, 2-1-piperazinyl ethanol, 2-hydroxyethylpiperazine, 2-piperazinoethanol, 2-piperazin-1-yl ethan-1-ol, 1-piperazinethanol, 4-2-hydroxyethyl piperazine
PubChem CID 7677
IUPAC Name 2-piperazin-1-ylethanol
SMILES C1CN(CCN1)CCO

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point -38.5°C
Color Colorless to Yellow
Density 1.0600g/mL
Boiling Point 245°C to 246°C
Flash Point 135°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Refractive Index 1.5055 to 1.5075
Quantity 100 g
Beilstein 23, II, 6
Specific Gravity 1.06
Solubility Information Solubility in water: miscible with water. Other solubilities: miscible with most common organic solvents
Formula Weight 130.19
Percent Purity 98.50%
Physical Form Oily Liquid
Chemical Name or Material N-(2-Hydroxyethyl)piperazine, 98.5%
Show More Show Less
compliance-icons
Product Identifier
  • N-(2-Hydroxyethyl)piperazine
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.